2-(2,2-Difluoroethoxy)-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

310535-1G 310535-500MG 310535-5G
77468-824EA 246.17 USD
77468-824 77481-506 77491-890
2-(2,2-Difluoroethoxy)-4-methylbenzaldehyde ≥95%
2-(2,2-Difluoroethoxy)-4-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₂
MW: 200.18 g/mol
Storage Temperature: Ambient
CAS Number: 2159867-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR