2-Fluoro-5-(methylsulfonyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-(methanesulphonyl)benzoic acid

30987-1G 30987-500MG 30987-5G
77468-820EA 88.58 USD
77468-820 77481-502 77491-886
2-Fluoro-5-(methylsulfonyl)benzoic acid ≥95%
2-Fluoro-5-(methylsulfonyl)benzoic acid

Precaución: For research use only

Formula: C₈H₇FO₄S
MW: 218.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11179900
CAS Number: 247569-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR