4-(Ethoxymethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

30973-1G 30973-500MG 30973-5G
77468-816EA 363.59 USD
77468-816 77481-498 77491-882
4-(Ethoxymethyl)benzaldehyde ≥95%
4-(Ethoxymethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20491959
CAS Number: 95068-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR