1-[2-Fluoro-3-(trifluoromethoxy)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

309680-1G 309680-500MG 309680-5G
77468-812EA 415.09 USD
77468-812 77481-494 77491-878
1-[2-Fluoro-3-(trifluoromethoxy)phenyl]-1-propanone ≥95%
1-[2-Fluoro-3-(trifluoromethoxy)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₀H₈F₄O₂
MW: 236.16 g/mol
Storage Temperature: Ambient
CAS Number: 1261838-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR