1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol 95

Supplier: AOBChem USA

Warning

309218-1G 309218-500MG 309218-5G
77468-804EA 415.09 USD
77468-804 77481-486 77491-870
1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol 95
1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₃ClF₄O
MW: 284.6776 g/mol
Storage Temperature: Ambient
CAS Number: 3002519-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR