(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

309215-1G 309215-500MG 309215-5G
77468-800EA 415.09 USD
77468-800 77481-482 77491-866
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₁ClF₄Si
MW: 270.7264 g/mol
Storage Temperature: Ambient
CAS Number: 3002511-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR