1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

309213-1G 309213-500MG 309213-5G
77468-796EA 415.09 USD
77468-796 77481-478 77491-862
1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-fluoroethanone ≥95%
1-(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₉H₄ClF₅O
MW: 258.5724 g/mol
Storage Temperature: Ambient
CAS Number: 3002434-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR