(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

309209-1G 309209-500MG 309209-5G
77468-792EA 415.09 USD
77468-792 77481-474 77491-858
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol ≥95%
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₉ClF₄O
Storage Temperature: Ambient
CAS Number: 3002485-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR