(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

309207-1G 309207-500MG 309207-5G
77468-788EA 415.09 USD
77468-788 77481-470 77491-854
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)triethylsilane ≥95%
(2-Chloro-6-fluoro-3-(trifluoromethyl)phenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₁₇ClF₄Si
MW: 312.8061 g/mol
Storage Temperature: Ambient
CAS Number: 3002541-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR