(3-(Benzyloxy)-6-bromo-2-fluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

309202-1G 309202-500MG 309202-5G
77468-784EA 415.09 USD
77468-784 77481-466 77491-850
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)triethylsilane ≥95%
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₉H₂₄BrFOSi
MW: 395.3811 g/mol
Storage Temperature: Ambient
CAS Number: 3002428-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR