(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

309200-1G 309200-500MG 309200-5G
77468-780EA 415.09 USD
77468-780 77481-462 77491-846
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(ethyl)sulfane ≥95%
(3-(Benzyloxy)-6-bromo-2-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₅H₁₄BrFOS
Storage Temperature: Ambient
CAS Number: 3002473-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR