2,6-Dibromo-4-methylbenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

30872-1G 30872-500MG 30872-5G
77468-772EA 397.58 USD
77468-772 77481-454 77491-838
2,6-Dibromo-4-methylbenzoic acid methyl ester ≥97%
2,6-Dibromo-4-methylbenzoic acid methyl ester

Precaución: For research use only

Formula: C₉H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29088220
CAS Number: 128733-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR