2-Bromo-6-chloro-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30804-1G 30804-500MG 30804-5G
77468-768EA 222.48 USD
77468-768 77481-450 77491-834
2-Bromo-6-chloro-3-methoxyphenylboronic acid pinacol ester ≥97%
Formula: C₁₃H₁₇BBrClO₃
Storage Temperature: Ambient
MDL Number: MFCD29088211
CAS Number: 2121513-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR