2-(Cyclopropylmethoxy)-3,5-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

307968-1G 307968-500MG 307968-5G
77468-764EA 676.71 USD
77468-764 77481-446 77491-830
2-(Cyclopropylmethoxy)-3,5-dimethylbenzoic acid ≥95%
2-(Cyclopropylmethoxy)-3,5-dimethylbenzoic acid

Precaución: For research use only

Formula: C₁₃H₁₆O₃
MW: 220.26 g/mol
Storage Temperature: Ambient
CAS Number: 1782318-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR