4-Chloro-2,3-difluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30792-1G 30792-500MG 30792-5G
77468-760EA 339.9 USD
77468-760 77481-442 77491-826
4-Chloro-2,3-difluoro-5-methoxybenzoic acid ≥97%
4-Chloro-2,3-difluoro-5-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₅ClF₂O₃
MW: 222.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088208
CAS Number: 1879026-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR