2,6-Dimethoxy-5-methylpyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

30757-1G 30757-500MG 30757-5G
77468-756EA 224.54 USD
77468-756 77481-438 77491-822
2,6-Dimethoxy-5-methylpyridine-3-carboxylic acid ≥95%
2,6-Dimethoxy-5-methylpyridine-3-carboxylic acid

Precaución: For research use only

Formula: C₉H₁₁NO₄
MW: 197.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088198
CAS Number: 1879026-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR