1-(2-Fluoro-5-propoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

306995-1G 306995-500MG 306995-5G
77468-752EA 415.09 USD
77468-752 77481-434 77491-818
1-(2-Fluoro-5-propoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Fluoro-5-propoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34892841
CAS Number: 2969329-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR