2-(2-Fluoro-5-propoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

306990-1G 306990-500MG 306990-5G
77468-748EA 415.09 USD
77468-748 77481-430 77491-814
2-(2-Fluoro-5-propoxyphenyl)propan-2-ol ≥95%
2-(2-Fluoro-5-propoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO₂
MW: 212.2605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892837
CAS Number: 2969452-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR