Cyclopropyl(2-fluoro-5-propoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

306988-1G 306988-500MG 306988-5G
77468-744EA 415.09 USD
77468-744 77481-426 77491-810
Cyclopropyl(2-fluoro-5-propoxyphenyl)methanol ≥95%
Cyclopropyl(2-fluoro-5-propoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₇FO₂
MW: 224.2712 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892835
CAS Number: 2969428-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR