2-Chloro-2,2-difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306986-1G 306986-500MG 306986-5G
77468-740EA 415.09 USD
77468-740 77481-422 77491-806
2-Chloro-2,2-difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₀ClF₃O₂
MW: 266.6441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892833
CAS Number: 2969476-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR