2,2-Difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306984-1G 306984-500MG 306984-5G
77468-736EA 415.09 USD
77468-736 77481-418 77491-802
2,2-Difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-fluoro-5-propoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁F₃O₂
MW: 232.1990 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892831
CAS Number: 2969405-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR