(2-Fluoro-5-propoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

306981-1G 306981-500MG 306981-5G
77468-732EA 415.09 USD
77468-732 77481-414 77491-798
(2-Fluoro-5-propoxyphenyl)trimethylsilane ≥95%
(2-Fluoro-5-propoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₁₉FOSi
MW: 226.3625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892828
CAS Number: 2969364-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR