2-Chloro-4-ethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30673-1G 30673-500MG 30673-5G
77468-728EA 82.4 USD
77468-728 77481-410 77491-794
2-Chloro-4-ethoxyphenylboronic acid pinacol ester ≥97%
2-Chloro-4-ethoxyphenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₄H₂₀BClO₃
MW: 282.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088173
CAS Number: 2121513-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR