1-(4-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306255-1G 306255-500MG 306255-5G
77468-724EA 415.09 USD
77468-724 77481-406 77491-788
1-(4-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone

Precaución: For research use only

Formula: C₁₀H₆BrFOS
MW: 273.1214 g/mol
Storage Temperature: Ambient
CAS Number: 3002533-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR