4-Bromo-2-(propylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

306252-1G 306252-500MG 306252-5G
77468-720EA 415.09 USD
77468-720 77481-402 77491-784
4-Bromo-2-(propylthio)benzo[b]thiophene ≥95%
4-Bromo-2-(propylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₁₁H₁₁BrS₂
Storage Temperature: Ambient
CAS Number: 3002466-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR