1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

306249-1G 306249-500MG 306249-5G
77468-716EA 415.09 USD
77468-716 77481-398 77491-780
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₁H₄BrF₅OS
MW: 359.1098 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR