4-Bromo-2-(ethylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

306247-1G 306247-500MG 306247-5G
77468-712EA 415.09 USD
77468-712 77481-394 77491-776
4-Bromo-2-(ethylthio)benzo[b]thiophene ≥95%
4-Bromo-2-(ethylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₁₀H₉BrS₂
Storage Temperature: Ambient
CAS Number: 3002471-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR