1-(4-Bromobenzo[b]thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

306245-1G 306245-500MG 306245-5G
77468-708EA 216.3 USD
77468-708 77481-390 77491-772
1-(4-Bromobenzo[b]thiophen-2-yl)ethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₁₀H₇BrOS
MW: 255.1309 g/mol
Storage Temperature: Ambient
CAS Number: 1895962-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR