2,4-Dibromobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

306243-1G 306243-500MG 306243-5G
77468-704EA 748.81 USD
77468-704 77481-386 77491-768
2,4-Dibromobenzo[b]thiophene ≥95%
2,4-Dibromobenzo[b]thiophene

Precaución: For research use only

Formula: C₈H₄Br₂S
Storage Temperature: Ambient
CAS Number: 1892558-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR