1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306241-1G 306241-500MG 306241-5G
77468-700EA 415.09 USD
77468-700 77481-382 77491-764
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(4-Bromobenzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₄BrF₃OS
Storage Temperature: Ambient
CAS Number: 3002525-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR