1-(5-Bromobenzofuran-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

306170-1G 306170-500MG 306170-5G
77468-696EA 415.09 USD
77468-696 77481-378 77491-760
1-(5-Bromobenzofuran-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(5-Bromobenzofuran-2-yl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₁H₄BrF₅O₂
MW: 343.0442 g/mol
Storage Temperature: Ambient
CAS Number: 3002473-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR