1-(5-Bromobenzofuran-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306167-1G 306167-500MG
77468-692EA 1610.92 USD
77468-692 77481-374
1-(5-Bromobenzofuran-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromobenzofuran-2-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₀H₄BrF₃O₂
MW: 293.0367 g/mol
Storage Temperature: Ambient
CAS Number: 1391766-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR