5-Bromo-2-(methylthio)benzofuran ≥95%

Supplier: AOBChem USA

Warning

306165-1G 306165-500MG 306165-5G
77468-688EA 415.09 USD
77468-688 77481-370 77491-754
5-Bromo-2-(methylthio)benzofuran ≥95%
5-Bromo-2-(methylthio)benzofuran

Precaución: For research use only

Formula: C₉H₇BrOS
Storage Temperature: Ambient
CAS Number: 3002487-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR