2-(Ethylthio)-5-methylbenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

306076-1G 306076-500MG 306076-5G
77468-680EA 415.09 USD
77468-680 77481-364 77491-746
2-(Ethylthio)-5-methylbenzo[b]thiophene ≥95%
2-(Ethylthio)-5-methylbenzo[b]thiophene

Precaución: For research use only

Formula: C₁₁H₁₂S₂
MW: 208.3429 g/mol
Storage Temperature: Ambient
CAS Number: 3002471-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR