1-(5-Bromobenzo[b]thiophen-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

306066-1G 306066-500MG 306066-5G
77468-672EA 415.09 USD
77468-672 77481-356 77491-738
1-(5-Bromobenzo[b]thiophen-2-yl)cyclobutanol ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
Storage Temperature: Ambient
CAS Number: 3002452-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR