(5-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

306043-1G 306043-500MG 306043-5G
77468-668EA 415.09 USD
77468-668 77481-352 77491-734
(5-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%
(5-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
MW: 283.1841 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892577
CAS Number: 2969404-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR