(5-Bromobenzo[b]thiophen-2-yl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

306041-1G 306041-500MG 306041-5G
77468-664EA 415.09 USD
77468-664 77481-348 77491-730
(5-Bromobenzo[b]thiophen-2-yl)triethylsilane ≥95%
(5-Bromobenzo[b]thiophen-2-yl)triethylsilane

Precaución: For research use only

Formula: C₁₄H₁₉BrSSi
Storage Temperature: Ambient
MDL Number: MFCD34892575
CAS Number: 2969449-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR