1-(5-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

306039-1G 306039-500MG 306039-5G
77468-660EA 415.09 USD
77468-660 77481-344 77491-726
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₅BrF₂OS
MW: 291.1119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892573
CAS Number: 2969449-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR