Methyl(2-Methyl-5-propoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

306027-1G 306027-500MG 306027-5G
77468-656EA 415.09 USD
77468-656 77481-340 77491-722
Methyl(2-Methyl-5-propoxyphenyl)sulfane ≥95%
Methyl(2-Methyl-5-propoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.3091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892571
CAS Number: 2969206-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR