1-Iodo-4-isopropoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

30602-1G 30602-500MG 30602-5G
77468-652EA 397.58 USD
77468-652 77481-336 77491-718
1-Iodo-4-isopropoxy-2-methylbenzene ≥95%
1-Iodo-4-isopropoxy-2-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃IO
MW: 276.114 g/mol
Storage Temperature: Ambient
CAS Number: 111220-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR