Isopropyl(5-methoxy-2-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

306017-1G 306017-500MG 306017-5G
77468-648EA 415.09 USD
77468-648 77481-332 77491-714
Isopropyl(5-methoxy-2-methylphenyl)sulfane ≥95%
Isopropyl(5-methoxy-2-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34892563
CAS Number: 2969449-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR