2-Ethyl-1-(5-methoxy-2-methylphenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

306014-1G 306014-500MG 306014-5G
77468-644EA 415.09 USD
77468-644 77481-328 77491-710
2-Ethyl-1-(5-methoxy-2-methylphenyl)butan-1-ol ≥95%
2-Ethyl-1-(5-methoxy-2-methylphenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.3232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34892562
CAS Number: 2969206-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR