1-(5-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

306011-1G 306011-500MG 306011-5G
77468-640EA 347.11 USD
77468-640 77481-324 77491-706
1-(5-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Methoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
Storage Temperature: Ambient
CAS Number: 2296246-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR