2,2,3,3,3-pentafluoro-1-(5-methoxy-2-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

306009-1G 306009-500MG 306009-5G
77468-636EA 415.09 USD
77468-636 77481-320 77491-702
2,2,3,3,3-pentafluoro-1-(5-methoxy-2-methylphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(5-methoxy-2-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₉F₅O₂
Storage Temperature: Ambient
CAS Number: 3002508-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR