Ethyl 2,6-dimethoxypyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

30599-1G 30599-500MG 30599-5G
77468-632EA 224.54 USD
77468-632 77481-316 77491-698
Ethyl 2,6-dimethoxypyridine-3-carboxylate ≥97%
Ethyl 2,6-dimethoxypyridine-3-carboxylate

Precaución: For research use only

Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11616888
CAS Number: 562840-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR