2-Bromo-6-fluorobiphenyl ≥95%

Supplier: AOBChem USA

Warning

30589-1G 30589-500MG 30589-5G
77468-628EA 195.7 USD
77468-628 77481-312 77491-694
2-Bromo-6-fluorobiphenyl ≥95%
2-Bromo-6-fluorobiphenyl

Precaución: For research use only

Formula: C₁₂H₈BrF
MW: 251.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32185816
CAS Number: 1233402-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR