Methyl 2-(2-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-methoxy-α-oxobenzeneacetate

305274-1G 305274-500MG 305274-5G
77468-624EA 85.49 USD
77468-624 77481-308 77491-690
Methyl 2-(2-methoxyphenyl)-2-oxoacetate ≥95%
Methyl 2-(2-methoxyphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617598
CAS Number: 5180-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR