Methyl 3-methoxy-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

305270-1G 305270-500MG 305270-5G
77468-620EA 327.54 USD
77468-620 77481-304 77491-686
Methyl 3-methoxy-α-oxobenzeneacetate ≥95%
Methyl 3-methoxy-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD11617588
CAS Number: 83585-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR