1-Bromo-3-isopropyl-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

30518-1G 30518-500MG 30518-5G
77468-616EA 235.87 USD
77468-616 77481-300 77491-682
1-Bromo-3-isopropyl-5-methoxybenzene ≥97%
1-Bromo-3-isopropyl-5-methoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356547
CAS Number: 1204344-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR