Ethyl 7-methylbenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

304472-1G 304472-500MG 304472-5G
77468-612EA 591.22 USD
77468-612 77481-296 77491-678
Ethyl 7-methylbenzo[b]thiophene-2-carboxylate ≥95%
Ethyl 7-methylbenzo[b]thiophene-2-carboxylate

Precaución: For research use only

Formula: C₁₂H₁₂O₂S
Storage Temperature: Ambient
CAS Number: 1514197-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR